Assay Details
Gene Symbol: Gria2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039195.1 | NP_001034284.1 | 7 - 8 | 1458 | 130 | |
NM_001083806.1 | NP_001077275.1 | 7 - 8 | 1458 | 130 | ||
NM_013540.2 | NP_038568.2 | 7 - 8 | 1458 | 130 | ||
GenBank mRNA | AB109210.1 | - | 7 - 8 | 1051 | 130 | |
AB109211.1 | - | 7 - 8 | 1051 | 130 | ||
AB111957.1 | - | 7 - 8 | 1051 | 130 | ||
AK014389.1 | - | 7 - 8 | 1503 | 130 | ||
AK043490.1 | - | 7 - 8 | 1431 | 130 | ||
AK044574.1 | - | 7 - 8 | 1390 | 130 | ||
AK046861.1 | - | 7 - 8 | 1458 | 130 | ||
AK086614.1 | - | 7 - 8 | 1392 | 130 | ||
AK134572.1 | - | 7 - 8 | 1388 | 130 | ||
AK134772.1 | - | 6 - 7 | 1029 | 130 | ||
AK141525.1 | - | 7 - 8 | 1434 | 130 | ||
AK163941.1 | - | 7 - 8 | 1451 | 130 | ||
BC048248.1 | - | 7 - 8 | 1317 | 6494013 | 130 | |
BC066193.1 | - | 7 - 8 | 1246 | 130 | ||
X57498.1 | - | 7 - 8 | 1253 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
