Assay Details
Gene Symbol: Snx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153680.2 | NP_710147.1 | 13 - 14 | 1359 | 95 | |
GenBank mRNA | AK077650.1 | - | 13 - 14 | 1361 | 95 | |
AK077780.1 | - | 13 - 14 | 1361 | 95 | ||
AK156299.1 | - | 13 - 14 | 1410 | 95 | ||
BC023732.1 | - | 13 - 14 | 1375 | 5352355 | 95 | |
BC023925.1 | - | 8 - 9 | 822 | 95 | ||
BC026571.1 | - | 13 - 14 | 1359 | 4986268 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
