Assay Details
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027276.2 | NP_081552.2 | 11 - 12 | 1090 | 91 | |
GenBank mRNA | AK147938.1 | - | 11 - 12 | 1105 | 91 | |
AK167965.1 | - | 11 - 12 | 1105 | 91 | ||
AK168978.1 | - | 11 - 12 | 1077 | 91 | ||
AK176938.1 | - | 1 - 2 | 63 | 91 | ||
BC026606.1 | - | 11 - 12 | 1090 | 4976424 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
