Assay Details
Gene Symbol: C1qtnf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175425.4 | NP_780634.2 | 1 - 2 | 58 | 64 | |
GenBank mRNA | BC038308.1 | - | 1 - 2 | 58 | 64 | |
DQ002400.1 | - | 1 - 2 | 67 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the DQ002400.1, NM_175425.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
