Assay Details
Gene Symbol: Pcdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122758.1 | NP_001116230.1 | 1 - 2 | 4260 | 73 | |
GenBank mRNA | AK039737.1 | - | 1 - 2 | 582 | 73 | |
AK165115.1 | - | 1 - 2 | 2162 | 73 | ||
AY861419.1 | - | 1 - 2 | 470 | 73 | ||
AY861420.1 | - | 1 - 2 | 470 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
