Assay Details
Gene Symbol: Chrna9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081104.1 | NP_001074573.1 | 3 - 4 | 694 | 63 | |
GenBank mRNA | AK010496.1 | - | 3 - 4 | 693 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
