Assay Details
Gene Symbol: Ogdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081130.1 | NP_001074599.1 | 13 - 14 | 1853 | 68 | |
GenBank mRNA | BC157970.1 | - | 12 - 13 | 1653 | 9007292 | 68 |
BC157971.1 | - | 12 - 13 | 1653 | 9007293 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
