Assay Details
Gene Symbol: Smim19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012667.2 | NP_001012685.2 | 3 - 4 | 568 | 107 | |
NM_001146117.1 | NP_001139589.1 | 2 - 3 | 509 | 107 | ||
GenBank mRNA | AK131902.1 | - | 2 - 3 | 260 | 107 | |
AK133712.1 | - | 2 - 3 | 513 | 107 | ||
AK140957.1 | - | 2 - 3 | 299 | 107 | ||
AK218393.1 | - | 1 - 2 | 125 | 107 | ||
BB618223.1 | - | 2 - 3 | 509 | 107 | ||
BC048089.1 | - | 3 - 4 | 544 | 6466443 | 107 | |
BQ922100.1 | - | 3 - 4 | 542 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
