Assay Details
Gene Symbol: Tbc1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289514.1 | NP_001276443.1 | 13 - 14 | 2141 | 124 | |
NM_019636.3 | NP_062610.2 | 14 - 15 | 2722 | 124 | ||
GenBank mRNA | AK122445.1 | - | 16 - 17 | 3810 | 124 | |
AK148356.1 | - | 13 - 14 | 2098 | 124 | ||
AK154591.1 | - | 15 - 16 | 2758 | 124 | ||
AK164923.1 | - | 10 - 11 | 1426 | 124 | ||
AK168763.1 | - | 3 - 4 | 427 | 124 | ||
BC004675.1 | - | 3 - 4 | 380 | 124 | ||
BC059824.1 | - | 14 - 15 | 2896 | 124 | ||
BC117931.1 | - | 13 - 14 | 2390 | 40097712 | 124 | |
U33005.1 | - | 14 - 15 | 2678 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
