Assay Details
Gene Symbol: Aco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007386.2 | NP_031412.2 | 16 - 17 | 2266 | 84 | |
GenBank mRNA | AK085951.1 | - | 16 - 17 | 2016 | 84 | |
AK129025.1 | - | 16 - 17 | 2048 | 84 | ||
AK163985.1 | - | 16 - 17 | 2065 | 84 | ||
X61147.1 | - | 16 - 17 | 2033 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
