Assay Details
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008628.2 | NP_032654.1 | 12 - 13 | 2039 | 136 | |
GenBank mRNA | AK157842.1 | - | 12 - 13 | 2052 | 136 | |
AK166179.1 | - | 12 - 13 | 2028 | 137 | ||
BC047117.1 | - | 12 - 13 | 2047 | 4951173 | 136 | |
BC050897.1 | - | 12 - 13 | 2022 | 6401835 | 136 | |
U21011.1 | - | 12 - 13 | 2056 | 136 | ||
X81143.1 | - | 12 - 13 | 2039 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
