Assay Details
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008628.2 | NP_032654.1 | 9 - 10 | 1552 | 87 | |
GenBank mRNA | AK051960.1 | - | 9 - 10 | 1549 | 87 | |
AK157842.1 | - | 9 - 10 | 1565 | 87 | ||
BC047117.1 | - | 9 - 10 | 1560 | 4951173 | 87 | |
BC050897.1 | - | 9 - 10 | 1535 | 6401835 | 87 | |
U21011.1 | - | 9 - 10 | 1569 | 87 | ||
X81143.1 | - | 9 - 10 | 1552 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
