Assay Details
Gene Symbol: Pah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008777.3 | NP_032803.2 | 2 - 3 | 389 | 68 | |
GenBank mRNA | AK149519.1 | - | 2 - 3 | 255 | 68 | |
BC013458.1 | - | 2 - 3 | 247 | 4223155 | 68 | |
X51942.1 | - | 2 - 3 | 214 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
