Assay Details
Gene Symbol: Kif5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008448.3 | NP_032474.2 | 19 - 20 | 2620 | 63 | |
GenBank mRNA | BC090841.1 | - | 19 - 20 | 2596 | 30543821 | 63 |
L27153.1 | - | 19 - 20 | 2214 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
