Assay Details
Gene Symbol: Trpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177781.4 | NP_808449.1 | 15 - 16 | 1927 | 91 | |
GenBank mRNA | AK045771.1 | - | 15 - 16 | 1928 | 91 | |
AY231177.1 | - | 15 - 16 | 1900 | 91 | ||
BC120563.1 | - | 15 - 16 | 1918 | 40129486 | 91 | |
BC131963.1 | - | 15 - 16 | 1918 | 40130240 | 91 | |
KF460461.1 | - | 15 - 16 | 1900 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
