Assay Details
Gene Symbol: Fbxo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081034.1 | NP_001074503.1 | 6 - 7 | 829 | 96 | |
GenBank mRNA | BC110686.1 | - | 6 - 7 | 568 | 96 | |
BC117884.1 | - | 6 - 7 | 575 | 40096054 | 96 | |
BC128479.1 | - | 6 - 7 | 579 | 40096053 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
