Assay Details
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019570.3 | NP_062516.2 | 1 - 2 | 205 | 74 | |
GenBank mRNA | AK048838.1 | - | 1 - 2 | 210 | 74 | |
BC058093.1 | - | 1 - 2 | 205 | 6827108 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC058093.1, NM_019570.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
