Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 11 - 12 | 1926 | 69 | |
GenBank mRNA | AB299229.1 | - | 11 - 12 | 1853 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
