Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 15 - 16 | 2569 | 82 | |
GenBank mRNA | AB299229.1 | - | 15 - 16 | 2496 | 82 | |
BC047418.1 | - | 5 - 6 | 682 | 82 | ||
BC051073.1 | - | 2 - 3 | 295 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
