Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 21 - 22 | 3372 | 79 | |
GenBank mRNA | AB299229.1 | - | 21 - 22 | 3299 | 79 | |
AF022178.1 | - | 6 - 7 | 687 | 79 | ||
AK050691.1 | - | 6 - 7 | 743 | 79 | ||
BC047418.1 | - | 11 - 12 | 1485 | 79 | ||
BC051073.1 | - | 8 - 9 | 1098 | 79 | ||
BC094568.1 | - | 3 - 4 | 656 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
