Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 22 - 23 | 3559 | 75 | |
GenBank mRNA | AB299229.1 | - | 22 - 23 | 3486 | 75 | |
AF022178.1 | - | 7 - 8 | 874 | 75 | ||
AK046668.1 | - | 1 - 2 | 48 | 75 | ||
AK050691.1 | - | 7 - 8 | 930 | 75 | ||
BC047418.1 | - | 12 - 13 | 1672 | 75 | ||
BC051073.1 | - | 9 - 10 | 1285 | 75 | ||
BC094568.1 | - | 4 - 5 | 843 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
