Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 27 - 28 | 4357 | 138 | |
GenBank mRNA | AB299229.1 | - | 27 - 28 | 4284 | 138 | |
AF081117.1 | - | 5 - 6 | 677 | 138 | ||
AK046668.1 | - | 6 - 7 | 846 | 138 | ||
BC047418.1 | - | 17 - 18 | 2470 | 138 | ||
BC094568.1 | - | 9 - 10 | 1641 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
