Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 28 - 29 | 4529 | 123 | |
GenBank mRNA | AB299229.1 | - | 28 - 29 | 4456 | 123 | |
AK046668.1 | - | 7 - 8 | 1018 | 123 | ||
BC047418.1 | - | 18 - 19 | 2642 | 123 | ||
BC094568.1 | - | 10 - 11 | 1813 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
