Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 4 - 5 | 623 | 81 | |
GenBank mRNA | AB299229.1 | - | 4 - 5 | 550 | 81 | |
AK143571.1 | - | 4 - 5 | 600 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
