Assay Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290729.1 | NP_001277658.1 | 5 - 6 | 865 | 89 | |
GenBank mRNA | AB299229.1 | - | 5 - 6 | 792 | 89 | |
AK045586.2 | - | 5 - 6 | 801 | 89 | ||
AK143571.1 | - | 5 - 6 | 842 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
