Assay Details
Gene Symbol: Scyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198021.2 | NP_932138.1 | 16 - 17 | 2304 | 81 | |
GenBank mRNA | AK136092.1 | - | 1 - 2 | 40 | 81 | |
AK139747.1 | - | 8 - 9 | 890 | 81 | ||
AK139761.1 | - | 16 - 17 | 2288 | 81 | ||
BC042443.1 | - | 16 - 17 | 2304 | 4037878 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
