Assay Details
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164634.1 | NP_001158106.1 | 4 - 5 | 823 | 94 | |
NM_001164635.1 | NP_001158107.1 | 4 - 5 | 731 | 94 | ||
NM_031194.5 | NP_112471.3 | 5 - 6 | 903 | 94 | ||
GenBank mRNA | AB079895.1 | - | 3 - 4 | 560 | 94 | |
AF078869.1 | - | 4 - 5 | 678 | 94 | ||
AK031962.1 | - | 5 - 6 | 869 | 94 | ||
AK044336.1 | - | 4 - 5 | 700 | 94 | ||
AK157891.1 | - | 4 - 5 | 732 | 94 | ||
BC014762.1 | - | 4 - 5 | 709 | 4239544 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
