Assay Details
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073190.1 | 2 - 3 | 753 | 87 | ||
GenBank mRNA | AK005057.1 | - | 2 - 3 | 754 | 87 | |
AK014174.1 | - | 2 - 3 | 716 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
