Assay Details
Gene Symbol: Socs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK048342.1 | - | 2 - 3 | 250 | 121 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 30510 |
Gm30510 | -13.4kb | Anti-sense | Chr. 17 - 87091755 - 87107347 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA C330024C12 gene | C330024C12Rik | 71.0kb | Anti-sense | Chr. 17 - 87192216 - 87194942 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
