Assay Details
Gene Symbol: Sf3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030109.2 | NP_084385.2 | 7 - 8 | 882 | 61 | |
GenBank mRNA | AK011613.1 | - | 7 - 8 | 841 | 61 | |
AK012009.1 | - | 7 - 8 | 836 | 61 | ||
AK012643.1 | - | 7 - 8 | 850 | 61 | ||
AK019282.1 | - | 7 - 8 | 828 | 61 | ||
AK142136.1 | - | 7 - 8 | 852 | 61 | ||
AK146063.1 | - | 7 - 8 | 852 | 61 | ||
AK146540.1 | - | 7 - 8 | 847 | 61 | ||
AK146864.1 | - | 7 - 8 | 851 | 61 | ||
AK147687.1 | - | 7 - 8 | 850 | 61 | ||
AK149776.1 | - | 7 - 8 | 851 | 61 | ||
AK150961.1 | - | 7 - 8 | 839 | 61 | ||
AK151354.1 | - | 7 - 8 | 829 | 61 | ||
AK152965.1 | - | 7 - 8 | 839 | 61 | ||
BC029745.1 | - | 7 - 8 | 817 | 61 | ||
BC045207.1 | - | 7 - 8 | 851 | 61 | ||
BC049118.1 | - | 7 - 8 | 875 | 6812422 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
