Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000019726.mm.1 |
TC1000002616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019726.mm.1 |
NM_013722.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019726.mm.1 |
NM_013722.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019723.mm.1 |
TC1000002616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019723.mm.1 |
NM_013722.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019723.mm.1 |
NM_013722.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019725.mm.1 |
TC1000002616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019725.mm.1 |
NM_013722.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019725.mm.1 |
NM_013722.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019727.mm.1 |
TC1000002616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019727.mm.1 |
NM_013722.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019727.mm.1 |
NM_013722.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019728.mm.1 |
TC1000002616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019728.mm.1 |
NM_013722.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019728.mm.1 |
NM_013722.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019722.mm.1 |
TC1000002616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019722.mm.1 |
NM_013722.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019722.mm.1 |
NM_013722.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019729.mm.1 |
TC1000002616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019729.mm.1 |
NM_013722.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019729.mm.1 |
NM_013722.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019730.mm.1 |
TC1000002616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019730.mm.1 |
NM_013722.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019730.mm.1 |
NM_013722.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019721.mm.1 |
TC1000002616.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019721.mm.1 |
NM_013722.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019721.mm.1 |
NM_013722.3 |
Refseq |