Assay Details
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028304.1 | - | 1 - 2 | 278 | 94 | |
AK088814.1 | - | 1 - 2 | 269 | 94 | ||
AK142658.1 | - | 1 - 2 | 269 | 94 | ||
AK145291.1 | - | 1 - 2 | 264 | 94 | ||
AK146207.1 | - | 1 - 2 | 268 | 94 | ||
AK150551.1 | - | 1 - 2 | 275 | 94 | ||
AK150777.1 | - | 1 - 2 | 233 | 94 | ||
AK151240.1 | - | 1 - 2 | 232 | 94 | ||
AK151409.1 | - | 1 - 2 | 275 | 94 | ||
AK151664.1 | - | 1 - 2 | 275 | 94 | ||
AK152338.1 | - | 1 - 2 | 275 | 94 | ||
AK152343.1 | - | 1 - 2 | 234 | 94 | ||
AK152991.1 | - | 1 - 2 | 234 | 94 | ||
AK153379.1 | - | 1 - 2 | 275 | 94 | ||
AK155905.1 | - | 1 - 2 | 269 | 94 | ||
AK159203.1 | - | 1 - 2 | 269 | 94 | ||
AK159350.1 | - | 1 - 2 | 269 | 94 | ||
AK159878.1 | - | 1 - 2 | 278 | 94 | ||
AK168708.1 | - | 1 - 2 | 211 | 94 | ||
AY029764.1 | - | 1 - 2 | 161 | 94 | ||
BC012401.1 | - | 1 - 2 | 258 | 3967323 | 94 | |
CT010238.1 | - | 1 - 2 | 126 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
