Assay Details
Gene Symbol: Gm34184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035016.1 | - | 4 - 5 | 491 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1500009L16 gene | 1500009L16Rik |
2700050C19Rik, OCC-1 |
-273.8kb | Anti-sense | Chr. 10 - 83722666 - 83762762 on NCBI Build GRCm38 |
Downstream |
NUAK family, SNF1-like kinase, 1 |
Nuak1 |
AU014801, AW494241, B230104P22Rik, Omphk1 |
327.2kb | Sense | Chr. 10 - 84370900 - 84440471 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
