Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700011553.mm.1 |
TC1700001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011553.mm.1 |
NM_001171052.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011553.mm.1 |
NM_001171052.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011553.mm.1 |
NM_001171053.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011553.mm.1 |
NM_001171053.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011553.mm.1 |
NM_001171054.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011553.mm.1 |
NM_001171054.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011553.mm.1 |
NM_054082.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011553.mm.1 |
NM_054082.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011552.mm.1 |
TC1700001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011552.mm.1 |
NM_001171052.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011552.mm.1 |
NM_001171052.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011552.mm.1 |
NM_001171053.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011552.mm.1 |
NM_001171053.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011552.mm.1 |
NM_001171054.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011552.mm.1 |
NM_001171054.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011552.mm.1 |
NM_054082.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011552.mm.1 |
NM_054082.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011554.mm.1 |
TC1700001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011554.mm.1 |
NM_001171052.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011554.mm.1 |
NM_001171052.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011554.mm.1 |
NM_001171053.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011554.mm.1 |
NM_001171053.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011554.mm.1 |
NM_001171054.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011554.mm.1 |
NM_001171054.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011554.mm.1 |
NM_054082.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011554.mm.1 |
NM_054082.2 |
Refseq |