Assay Details
Gene Symbol: Mib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144860.2 | NP_659109.2 | 4 - 5 | 1314 | 73 | |
GenBank mRNA | AK139338.1 | - | 4 - 5 | 1195 | 73 | |
AY147848.1 | - | 4 - 5 | 813 | 73 | ||
AY149907.1 | - | 4 - 5 | 634 | 73 | ||
AY245539.1 | - | 4 - 5 | 1314 | 73 | ||
AY974091.1 | - | 4 - 5 | 607 | 73 | ||
BC131662.1 | - | 5 - 6 | 780 | 40110696 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
