Assay Details
Gene Symbol: Mib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144860.2 | NP_659109.2 | 7 - 8 | 1768 | 99 | |
GenBank mRNA | AK080847.1 | - | 1 - 2 | 44 | 99 | |
AY147848.1 | - | 7 - 8 | 1267 | 99 | ||
AY149907.1 | - | 7 - 8 | 1088 | 99 | ||
AY245539.1 | - | 7 - 8 | 1768 | 99 | ||
AY974091.1 | - | 7 - 8 | 1061 | 99 | ||
BC131662.1 | - | 8 - 9 | 1234 | 40110696 | 99 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK080847.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
