Assay Details
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011163.4 | NP_035293.1 | 6 - 7 | 764 | 82 | |
GenBank mRNA | AK028602.1 | - | 6 - 7 | 764 | 82 | |
BC016422.1 | - | 6 - 7 | 727 | 3964935 | 82 | |
M65029.1 | - | 6 - 7 | 716 | 82 | ||
M93567.1 | - | 6 - 7 | 706 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
