Assay Details
Gene Symbol: Pik3r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081309.1 | NP_001074778.1 | 6 - 7 | 2244 | 60 | |
GenBank mRNA | AK050333.1 | - | 6 - 7 | 2214 | 60 | |
AK087938.1 | - | 6 - 7 | 2235 | 60 | ||
AK169508.1 | - | 5 - 6 | 3031 | 60 | ||
BC017537.1 | - | 5 - 6 | 1846 | 60 | ||
BC024083.1 | - | 3 - 4 | 718 | 60 | ||
BC157947.1 | - | 6 - 7 | 2069 | 9007281 | 60 | |
BC158047.1 | - | 6 - 7 | 2069 | 9088160 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
