Assay Details
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110791.1 | NP_001104261.1 | 3 - 4 | 632 | 135 | |
NM_011750.2 | NP_035880.2 | 3 - 4 | 632 | 135 | ||
GenBank mRNA | AK133895.1 | - | 3 - 4 | 421 | 135 | |
AK146151.1 | - | 3 - 4 | 393 | 135 | ||
AK147081.1 | - | 3 - 4 | 401 | 135 | ||
AK154705.1 | - | 3 - 4 | 401 | 135 | ||
AK157845.1 | - | 3 - 4 | 401 | 135 | ||
BC009091.1 | - | 3 - 4 | 377 | 3157495 | 135 | |
BC055370.1 | - | 3 - 4 | 391 | 30055109 | 135 | |
X85802.1 | - | 3 - 4 | 389 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
