Assay Details
Gene Symbol: Lonp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028782.2 | NP_083058.2 | 11 - 12 | 1776 | 65 | |
GenBank mRNA | AK004820.1 | - | 11 - 12 | 1760 | 65 | |
AK131910.1 | - | 11 - 12 | 1762 | 65 | ||
AK157474.1 | - | 11 - 12 | 1755 | 65 | ||
AK159302.1 | - | 11 - 12 | 1761 | 65 | ||
AK160071.1 | - | 11 - 12 | 1761 | 65 | ||
AK160076.1 | - | 11 - 12 | 1762 | 65 | ||
AK161983.1 | - | 11 - 12 | 1761 | 65 | ||
AY190302.1 | - | 11 - 12 | 1746 | 65 | ||
BC057365.1 | - | 9 - 10 | 3170 | 65 | ||
BC058735.1 | - | 9 - 10 | 3170 | 65 | ||
BC092212.1 | - | 2 - 3 | 143 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
