Assay Details
Gene Symbol: Galnt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027864.2 | NP_082140.2 | 6 - 7 | 1042 | 64 | |
GenBank mRNA | AB175681.1 | - | 6 - 7 | 650 | 64 | |
AK002747.1 | - | 6 - 7 | 1014 | 64 | ||
AK049598.1 | - | 6 - 7 | 988 | 64 | ||
AK078292.1 | - | 6 - 7 | 655 | 64 | ||
BC117800.1 | - | 6 - 7 | 826 | 40092267 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
