Assay Details
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK133647.1 | - | 2450 | 116 | ||
M37890.1 | - | 6 - 7 | 2389 | 116 | ||
S56585.1 | - | 6 - 7 | 2468 | 116 | ||
X53779.1 | - | 6 - 7 | 2421 | 116 | ||
X59411.1 | - | 6 - 7 | 2420 | 116 | ||
X59592.1 | - | 6 - 7 | 2509 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
