Assay Details
Gene Symbol: Nek11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172461.3 | NP_766049.2 | 9 - 10 | 1193 | 59 | |
GenBank mRNA | AK030042.1 | - | 9 - 10 | 1152 | 59 | |
AK030186.1 | - | 8 - 9 | 1069 | 59 | ||
AK043543.1 | - | 9 - 10 | 1432 | 59 | ||
AK054237.1 | - | 9 - 10 | 1087 | 59 | ||
BC119588.1 | - | 8 - 9 | 1027 | 40058889 | 59 | |
BC119589.1 | - | 8 - 9 | 1027 | 40058890 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
