Assay Details
Gene Symbol: Tbc1d8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018775.4 | NP_061245.3 | 2 - 3 | 422 | 61 | |
GenBank mRNA | AK138630.1 | - | 2 - 3 | 420 | 61 | |
AK141520.1 | - | 2 - 3 | 411 | 61 | ||
AK141816.1 | - | 2 - 3 | 376 | 61 | ||
BC065081.1 | - | 2 - 3 | 356 | 6832495 | 61 | |
CJ139650.1 | - | 2 - 3 | 423 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
