Assay Details
Gene Symbol: Copb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015827.2 | NP_056642.1 | 2 - 3 | 229 | 76 | |
GenBank mRNA | AK088064.1 | - | 2 - 3 | 230 | 76 | |
AK153352.1 | - | 2 - 3 | 230 | 76 | ||
BC006675.1 | - | 2 - 3 | 216 | 3499585 | 76 | |
BC010842.1 | - | 2 - 3 | 219 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
