Assay Details
Gene Symbol: Cdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285988.1 | NP_001272917.1 | 13 - 14 | 1951 | 106 | |
NM_001285989.1 | NP_001272918.1 | 13 - 14 | 1933 | 106 | ||
NM_001285990.1 | NP_001272919.1 | 12 - 13 | 1856 | 106 | ||
NM_145436.2 | NP_663411.2 | 13 - 14 | 1936 | 106 | ||
GenBank mRNA | AK148278.1 | - | 13 - 14 | 1791 | 106 | |
AK161143.1 | - | 2 - 3 | 312 | 106 | ||
AK161471.1 | - | 13 - 14 | 1802 | 106 | ||
BC157955.1 | - | 13 - 14 | 1950 | 9088128 | 106 | |
BC172100.1 | - | 13 - 14 | 1950 | 9054386 | 106 | |
BC172149.1 | - | 12 - 13 | 1855 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
