Assay Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168591.1 | NP_001162063.1 | 7 - 8 | 976 | 90 | |
NM_025827.3 | NP_080103.1 | 13 - 14 | 2280 | 90 | ||
GenBank mRNA | AK003766.1 | - | 1 - 2 | 117 | 90 | |
AK004843.1 | - | 13 - 14 | 2194 | 90 | ||
AK050231.1 | - | 13 - 14 | 2189 | 90 | ||
AK075719.1 | - | 7 - 8 | 977 | 90 | ||
AK088348.1 | - | 12 - 13 | 2186 | 90 | ||
AK151103.1 | - | 13 - 14 | 2226 | 90 | ||
AK151406.1 | - | 11 - 12 | 1794 | 90 | ||
AK152964.1 | - | 11 - 12 | 1796 | 90 | ||
AK168628.1 | - | 11 - 12 | 1804 | 90 | ||
AK168791.1 | - | 13 - 14 | 2226 | 90 | ||
AK168956.1 | - | 13 - 14 | 2226 | 90 | ||
AK211044.1 | - | 1 - 2 | 68 | 90 | ||
BC049090.1 | - | 13 - 14 | 2216 | 6401542 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
