Assay Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025827.3 | NP_080103.1 | 2 - 3 | 605 | 69 | |
GenBank mRNA | AK004843.1 | - | 2 - 3 | 519 | 69 | |
AK050231.1 | - | 2 - 3 | 514 | 69 | ||
AK088348.1 | - | 2 - 3 | 606 | 69 | ||
AK151103.1 | - | 2 - 3 | 551 | 69 | ||
AK151406.1 | - | 2 - 3 | 545 | 69 | ||
AK152964.1 | - | 2 - 3 | 545 | 69 | ||
AK168628.1 | - | 2 - 3 | 555 | 69 | ||
AK168791.1 | - | 2 - 3 | 551 | 69 | ||
AK168956.1 | - | 2 - 3 | 551 | 69 | ||
AK169656.1 | - | 519 | 69 | |||
BC049090.1 | - | 2 - 3 | 541 | 6401542 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
