Assay Details
Gene Symbol: Gria4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113180.1 | NP_001106651.1 | 13 - 14 | 2525 | 115 | |
NM_019691.4 | NP_062665.3 | 13 - 14 | 2525 | 115 | ||
GenBank mRNA | AB022913.1 | - | 13 - 14 | 2259 | 115 | |
AB079073.1 | - | 12 - 13 | 2046 | 115 | ||
AB104708.1 | - | 2 - 3 | 410 | 115 | ||
AK031568.1 | - | 13 - 14 | 2499 | 115 | ||
AK036193.1 | - | 2 - 3 | 408 | 115 | ||
AK135034.1 | - | 13 - 14 | 2232 | 115 | ||
BC060697.1 | - | 13 - 14 | 2433 | 6409918 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
