Assay Details
Gene Symbol: Gria4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113180.1 | NP_001106651.1 | 1 - 2 | 396 | 90 | |
NM_001113181.1 | NP_001106652.1 | 1 - 2 | 396 | 90 | ||
NM_019691.4 | NP_062665.3 | 1 - 2 | 396 | 90 | ||
GenBank mRNA | AK031568.1 | - | 1 - 2 | 369 | 90 | |
AK032103.1 | - | 1 - 2 | 397 | 90 | ||
AK135034.1 | - | 1 - 2 | 102 | 90 | ||
BC060697.1 | - | 1 - 2 | 304 | 6409918 | 90 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK031568.1, AK032103.1, BC060697.1, NM_001113180.1, NM_001113181.1, NM_019691.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
